Limited time offer: 3 year warranty when you purchase a SimpliAmp Thermal Cycler. View flyer »

Pierce™ Streptavidin, Hydrazide-Activated

Catalog number: 21120B

Thermo Scientific™  Related applications: Protein Labeling & Crosslinking

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce Hydrazide-Activated Streptavidin conjugate include recombinant streptavidin in a purified form activated for crosslinking to carbonyl groups in a molecule.

Related Products
Pierce™ Streptavidin
Pierce™ Streptavidin, Horseradish Peroxidase Conjugated
Pierce™ Streptavidin, Alkaline Phosphatase Conjugated
Pierce™ Streptavidin, Maleimide-Activated
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity: Aldehyde, Ketone
Label or Dye: Streptavidin
Product Line: Pierce™
Product Size: Custom
Solubility: Water

Contents & storage

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.

Documents

Manuals & protocols