Xylenol Orange, sodium salt, pure, water soluble
Xylenol Orange, sodium salt, pure, water soluble
Xylenol Orange, sodium salt, pure, water soluble
Thermo Scientific Chemicals

Xylenol Orange, sodium salt, pure, water soluble

CAS: 3618-43-7 | C31H28N2Na4O13S
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 211220010
Price (EUR)
47,60
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
47,60
Each
Chemical Identifiers
CAS3618-43-7
IUPAC Nametetrasodium 2-[({5-[3-(3-{[bis(carboxylatomethyl)amino]methyl}-4-hydroxy-5-methylphenyl)-2,2-dioxo-3H-1,2λ⁶-benzoxathiol-3-yl]-2-hydroxy-3-methylphenyl}methyl)(carboxylatomethyl)amino]acetate
Molecular FormulaC31H28N2Na4O13S
InChI KeyFXYMNKLSOQONJB-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].CC1=CC(=CC(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(C2=CC=CC=C2OS1(=O)=O)C1=CC(C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Loss on drying=<10.0 % (110°C, 1 h)
Appearance (Form)Crystalline powder
UV spectrumComplies
Appearance (Color)Red-brown to dark brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only