Silver chloride, 99+%, extra pure
Silver chloride, 99+%, extra pure
Silver chloride, 99+%, extra pure
Thermo Scientific Chemicals

Silver chloride, 99+%, extra pure

CAS: 7783-90-6 | AgCl | 143.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 211272500
Price (USD)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7783-90-6
IUPAC Namesilver(1+) chloride
Molecular FormulaAgCl
InChI KeyHKZLPVFGJNLROG-UHFFFAOYSA-M
SMILES[Cl-].[Ag+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pink-beige to grey
Iron (Fe)=<10 ppm
Lead (Pb)=<10 ppm
Titration>=99.0 %
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only