Pierce™ Avidin
Thermo Scientific™

Pierce™ Avidin

Thermo Scientific Pierce Avidin is a purified avidin protein from hen egg whites that effectively binds biotin, a tag thatRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2112820 mg
2112110 mg
Catalog number 21128
Price (USD)
258.00
Each
Quantity:
20 mg
Price (USD)
258.00
Each
Thermo Scientific Pierce Avidin is a purified avidin protein from hen egg whites that effectively binds biotin, a tag that is frequently used to tag antibodies and other probes for immunodetection methods.

Features of Pierce Avidin:

Native avidin—glycoprotein from chicken egg whites, 67kDa, pI = 10
Soluble—glycosylation and isoelectric point renders avidin more soluble than streptavidin
Affordable—significantly less expensive than recombinant streptavidin
ABC staining—avidin is usually preferred over streptavidin for avidin-biotin complex (ABC) staining methods in immunohistochemistry (IHC)
Pure protein—choose pure, unconjugated avidin for use in ABC staining

Avidin is a glycoprotein found in the egg white and tissues of birds, reptiles and amphibia. This protein contains four identical subunits having a combined molecular mass of 67,000 to 68,000 daltons. Each subunit binds one molecule of biotin, and studies have shown that tryptophan and lysine are involved in the binding site for biotin. The sequencing of the subunit indicates it consists of 128 amino acids. Avidin has an isoelectric point of 10-10.5 and is very soluble in water and salt solutions. Avidin is stable over a wide range of pH values and temperatures. Extensive chemical modification has little effect on the activity of avidin, making it useful for detection and protein purification.

Related Products
Pierce™ Avidin, Horseradish Peroxidase Conjugated
Pierce™ Avidin, Fluorescein (FITC) Conjugated
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeUnlabeled
Product TypeAvidin (Unconjugated)
Quantity20 mg
Target MoleculeBiotin
ConjugateUnconjugated
FormSolid
Product LinePierce
Unit SizeEach
Contents & Storage
Store at -20°C. Shipped at ambient temperature.