Pierce™ Avidin

Catalog number: 21128B

Thermo Scientific™  Related applications: Cellular Imaging | Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Thermo Scientific Pierce Avidin is a purified avidin protein from hen egg whites that effectively binds biotin, a tag that is frequently used to tag antibodies and other probes for immunodetection methods.

Features of Pierce Avidin:

Native avidin—glycoprotein from chicken egg whites, 67kDa, pI = 10
Soluble—glycosylation and isoelectric point renders avidin more soluble than streptavidin
Affordable—significantly less expensive than recombinant streptavidin
ABC staining—avidin is usually preferred over streptavidin for avidin-biotin complex (ABC) staining methods in immunohistochemistry (IHC)
Pure protein—choose pure, unconjugated avidin for use in ABC staining

Avidin is a glycoprotein found in the egg white and tissues of birds, reptiles and amphibia. This protein contains four identical subunits having a combined molecular mass of 67,000 to 68,000 daltons. Each subunit binds one molecule of biotin, and studies have shown that tryptophan and lysine are involved in the binding site for biotin. The sequencing of the subunit indicates it consists of 128 amino acids. Avidin has an isoelectric point of 10-10.5 and is very soluble in water and salt solutions. Avidin is stable over a wide range of pH values and temperatures. Extensive chemical modification has little effect on the activity of avidin, making it useful for detection and protein purification.

Related Products
Pierce™ Avidin, Horseradish Peroxidase Conjugated
Pierce™ Avidin, Fluorescein (FITC) Conjugated
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Label or Dye: Unlabeled
Product Line: Pierce™
Product Size: Custom
Target Molecule: Biotin

Contents & storage

Store at -20°C. Shipped at ambient temperature.

Documents

Manuals & protocols