Chloro(1,5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh
Chloro(1,5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh
Chloro(1,5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh
Thermo Scientific Chemicals

Chloro(1,5-cyclooctadiene)rhodium(I) dimer, min. 40.8% Rh

CAS: 12092-47-6 | C16H24Cl2Rh2 | 493.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Catalog number 211300050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS12092-47-6
IUPAC Namebis(λ¹-rhodium(1+)) bis((1Z,5Z)-cycloocta-1,5-diene) dichloride
Molecular FormulaC16H24Cl2Rh2
InChI KeyQSUDXYGZLAJAQU-MIXQCLKLSA-L
SMILES[Cl-].[Cl-].[Rh+].[Rh+].C1C\C=C/CC\C=C/1.C1C\C=C/CC\C=C/1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to orange-brown
Infrared spectrumConforms
Appearance (Form)Crystalline powder or crystals
Rhodium (Rh)>=40.8 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only