Potassium peroxomonosulfate, extra pure, min. 4.5% active oxygen
Potassium peroxomonosulfate, extra pure, min. 4.5% active oxygen
Potassium peroxomonosulfate, extra pure, min. 4.5% active oxygen
Thermo Scientific Chemicals

Potassium peroxomonosulfate, extra pure, min. 4.5% active oxygen

CAS: 70693-62-8 | H3K5O18S4 | 614.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 211361000
Price (EUR)
26,65
Online exclusive
29,90
Save 3,25
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
26,65
Online exclusive
29,90
Save 3,25
Each
Chemical Identifiers
CAS70693-62-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline or granular powder
Water=<0.5 %
Appearance (Color)White
Titration>=4.5 % (Active oxygen)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only