Neodymium(III)-oxide, 99.9%, (trace metal basis)
Neodymium(III)-oxide, 99.9%, (trace metal basis)
Neodymium(III)-oxide, 99.9%, (trace metal basis)
Thermo Scientific Chemicals

Neodymium(III)-oxide, 99.9%, (trace metal basis)

CAS: 1313-97-9 | Nd2O3 | 336.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 211580250
Price (EUR)
58,65
Online exclusive
65,40
Save 6,75 (10%)
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
58,65
Online exclusive
65,40
Save 6,75 (10%)
Each
Chemical Identifiers
CAS1313-97-9
IUPAC Namedineodymium(3+) trioxidandiide
Molecular FormulaNd2O3
InChI KeyPLDDOISOJJCEMH-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Nd+3].[Nd+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Blue
Total trace metal impurities=<0.1 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only