Zinc nitrate hexahydrate, 98%, extra pure
Zinc nitrate hexahydrate, 98%, extra pure
Zinc nitrate hexahydrate, 98%, extra pure
Zinc nitrate hexahydrate, 98%, extra pure
Acros Organics

Zinc nitrate hexahydrate, 98%, extra pure

CAS: 10196-18-6 | H12N2O12Zn | 297.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
1 kg
Packaging:
View all
Glass Bottle
Catalog number 211660050
HSN 28342990
Price (INR)
3,432.00
Each
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
3,432.00
Each
Add to cart
Chemical Identifiers
CAS10196-18-6
IUPAC Namezinc(2+) hexahydrate dinitrate
Molecular FormulaH12N2O12Zn
InChI KeyJGPSMWXKRPZZRG-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Zn++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Adhering crystals and/or chunks
Iron (Fe)=<100 ppm
Lead (Pb)=<10 ppm
Chloride (Cl)=<100 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only