Bacto™ Peptone
Gibco™

Bacto™ Peptone

Bacto Peptone is an animal-origin (AO) enzymatic digest of bovine and porcine animal proteins.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityFormat
211677500 gBottle
21183010 kgPail
2118202 kgPail
Catalog number 211677
Price (USD)
171.00
Each
Quantity:
500 g
Format:
Bottle
Price (USD)
171.00
Each
Bacto Peptone is an animal-origin enzymatic digest of bovine and porcine animal proteins. The nutritive value of Bacto Peptone is largely dependent on a combination of a negligible quantity of proteases, high concentration of complex constituents, and a unique amino acid content that makes this peptone a strong nitrogen source. Customers have successfully used this supplement with a variety of cell types including Saccharomyces cerevisiae, Escherichia coli, Pasteurella multocida, Vibrio cholerae, and porcine kidney cells to produce biotherapeutics and vaccines for animal health applications.

Features:

  • A strong nitrogen source
  • Unique amino acid content
  • High concentration of complex constituents

Applications of Bacto Peptone include:

  • Vaccines for animal health applications
  • Production of biotherapeutics
  • Mammalian cell culture
  • Microbial fermentation
Specifications
Cell TypeBacteria
Certifications/ComplianceLUO, MUO
Culture TypeMammalian, Insect, Microbial
For Use With (Application)Formulation of culture media for the cultivation of heterotrophic microorganisms
FormatBottle
Product LineBacto
Quantity500 g
Shelf Life1825 Days
SourceAnimal origin
FormPowder
Product TypeBacterial Media
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.