Nalgene™ Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closure
Nalgene™ Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closure
Nalgene™ Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closure
Nalgene™ Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closure
Thermo Scientific™

Nalgene™ Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closure

Protect against breakage and contamination with museum-quality PMP jars that lose only 2% of formaldehyde per year.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
2117-006060 mL53 mm
2117-0125125 mL70 mm
2117-0250250 mL70 mm
2117-0500500 mL120 mm
2117-10001000 mL120 mm
Catalog number 2117-0060
Price (BRL)
-
Capacity (Metric):
60 mL
Closure Size:
53 mm

Protect against breakage and contamination with Thermo Scientific™ Nalgene™; Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closure. These make excellent museum jars offering good clarity with excellent chemical resistance. They are lighter in weight than glass and lose only 2% of formaldehyde per year. Not liquid leakproof.

Features:
  • 125mL to 1L jars have white polypropylene closures with recessed tops for stacking.
  • Good clarity with excellent chemical resistance make these jars versatile in your lab.
  • Straight sides provide full-access and complete sample retrieval.
  • 125 and 250mL sizes fit Osterizer™ and other blenders that have Mason Jar threads.
  • They are not liquid leakproof, not recommended for liquid transport.
  • Chemically compatible with a variety of laboratory chemicals and reagents. Consult the Nalgene Chemical Resistance Guide for specific chemical compatibility information.
  • Autoclavable/Transparent 

Note: Completely disengage threads or remove cap before autoclaving.

Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene  products are packaged.

.

Specifications
Capacity2 oz., 60 mL
Capacity (English)2 oz.
ClarityClear
Closure MaterialPolypropylene
Closure Size53 mm
Closure TypeScrew Cap
Diameter (English)1.8 in.
Diameter (English) Inner Neck1.7 in.
Diameter (Metric)47 mm
Diameter (Metric) Inner Neck44 mm
Height (English)1.9 in.
Height (Metric)45 mm
IncludesScrew Closure
No. per Case48
No. per Pack4
Product TypeStraight-sided Jar
ShapeRound
Weight (Metric)26 g
Capacity (Metric)60 mL
ColorClear
MaterialPMP
MouthWide
Unit SizeCase of 48

Frequently asked questions (FAQs)

Why are some of your wide mouth bottles and jars not considered liquid leakproof?

This was a consequence of design and engineering limitations, and there was no practical way to incorporate the same level of seal integrity as our certified leakproof containers. While in many cases, they still hold up well to liquid containment applications, they have not been tested specifically for that purpose, and thus would be the responsibility of the customer to perform validation and risk management studies if necessary.