Nalgene™ Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closure
Nalgene™ Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closure
Nalgene™ Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closure
Nalgene™ Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closure
Thermo Scientific™

Nalgene™ Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closure

Protect against breakage and contamination with museum-quality PMP jars that lose only 2% of formaldehyde per year.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure SizeUnit Size
2117-0500PK
also known as 2117-0500
500 mL120 mmPack of 4
2117-006060 mL53 mmCase of 48
2117-0060PK
also known as 2117-0060
60 mL53 mmPack of 4
2117-0125125 mL70 mmCase of 24
2117-0125PK
also known as 2117-0125
125 mL70 mmPack of 4
2117-0250250 mL70 mmCase of 24
2117-0250PK
also known as 2117-0250
250 mL70 mmPack of 4
2117-0500500 mL120 mmCase of 16
2117-10001000 mL120 mmCase of 16
2117-1000PK
also known as 2117-1000
1000 mL120 mmPack of 4
Catalog number 2117-0500PK
also known as 2117-0500
Price (HKD)
-
Capacity (Metric):
500 mL
Closure Size:
120 mm
Unit Size:
Pack of 4

Protect against breakage and contamination with Thermo Scientific™ Nalgene™; Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closure. These make excellent museum jars offering good clarity with excellent chemical resistance. They are lighter in weight than glass and lose only 2% of formaldehyde per year. Not liquid leakproof.

Features:
  • 125mL to 1L jars have white polypropylene closures with recessed tops for stacking.
  • Good clarity with excellent chemical resistance make these jars versatile in your lab.
  • Straight sides provide full-access and complete sample retrieval.
  • 125 and 250mL sizes fit Osterizer™ and other blenders that have Mason Jar threads.
  • They are not liquid leakproof, not recommended for liquid transport.
  • Chemically compatible with a variety of laboratory chemicals and reagents. Consult the Nalgene Chemical Resistance Guide for specific chemical compatibility information.
  • Autoclavable/Transparent 

Note: Completely disengage threads or remove cap before autoclaving.

Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene  products are packaged.

.

Specifications
Capacity16 oz., 500 mL
Capacity (English)16 oz.
ClarityClear
Closure MaterialPolypropylene
Closure Size120 mm
Closure TypeScrew Cap
Diameter (English)4.4 in.
Diameter (English) Inner Neck4.4 in.
Diameter (Metric)112 mm
Diameter (Metric) Inner Neck112 mm
Height (English)3.4 in.
Height (Metric)88 mm
IncludesScrew Closure
No. per Case16
No. per Pack4
Product TypeJar
ShapeRound
Weight (Metric)0.13 kg
Capacity (Metric)500 mL
ColorClear
MaterialPMP
MouthWide
Unit SizePack of 4

Frequently asked questions (FAQs)

Why are some of your wide mouth bottles and jars not considered liquid leakproof?

This was a consequence of design and engineering limitations, and there was no practical way to incorporate the same level of seal integrity as our certified leakproof containers. While in many cases, they still hold up well to liquid containment applications, they have not been tested specifically for that purpose, and thus would be the responsibility of the customer to perform validation and risk management studies if necessary.