L(+)-Tartaric acid diammonium salt, 99%
L(+)-Tartaric acid diammonium salt, 99%
L(+)-Tartaric acid diammonium salt, 99%
Thermo Scientific Chemicals

L(+)-Tartaric acid diammonium salt, 99%

CAS: 3164-29-2 | C4H12N2O6 | 184.15 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
產品號碼 211812500
價格 (TWD)
-
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS3164-29-2
IUPAC Name(2R,3R)-2,3-dihydroxybutanedioic acid diamine
Molecular FormulaC4H12N2O6
InChI KeyNGPGDYLVALNKEG-OLXYHTOASA-N
SMILESN.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
檢視更多
規格Specification Sheet規格表
Titration with HClO4>=98.5 %
Specific optical rotation+31.4° to +33.4° (20°C, 589 nm) (c=1.84, H2O)
Sulfate (SO4)=<50 ppm
Heavy metals (as Pb)=<10 ppm
Iron (Fe)=<10 ppm
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only