Pierce™ Recombinant Protein A

Catalog number: 21184B

Thermo Scientific™  Related applications: Cellular Imaging | Protein Assays and Analysis

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}} also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Purified (unconjugated) Thermo Scientific Pierce Recombinant Protein A is useful as the basis for preparing various kinds of probes or affinity media for detection or purification of rabbit and human antibodies, especially IgG isotypes, in immunoassays and antibody purification protocols.

Features of Recombinant Protein A:

• Contains four Fc-binding domains per protein
• Better than Protein G for polyclonal IgG from rabbit, pig, dog and cat serum
• Poorer than Protein G for mouse IgG1, human IgG3 and rat, goat and cow antibodies

Protein A is a cell wall component produced by several strains of Staphylococcus aureus that consists of a single polypeptide chain and contains little or no carbohydrate. Recombinant Protein A is produced in E. coli and functions essentially the same as native Protein A. The Protein A molecule contains four high-affinity binding sites capable of interacting with the Fc region from IgG of several species including human and rabbit. Optimal binding occurs at pH 8.2, although binding is also effective at neutral or physiological conditions (pH 7.0 to 7.6).

The interaction between Protein A and IgG is not equivalent for all species. Even within a species, Protein A interacts with some subclasses of IgG and not others. For instance, human IgG1, IgG2 and IgG4 bind strongly, while IgG3 does not bind. There are also many instances in which monoclonal antibodies do not bind to Protein A, especially the majority of rat immunoglobulins and mouse IgG1.

Properties of Recombinant Protein A:
• Source: E. coli
• Molecular Weight: ~44,600 (Apparent MW by SDS-PAGE: 45,000)
• Form: salt-free powder
• A280 of 0.1% solution: ~0.165
• Isoelectric point (pI): 4.7-4.8

Related Products
Pierce™ Recombinant Protein A, Peroxidase Conjugated
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Product Line: Pierce™
Product Size: Custom
Target Molecule: Antibodies

Contents & storage

Upon receipt store at -20°C. Product is shipped at ambient temperature.

Documents

Manuals & protocols