Keep your oxygen scavenger analyzer ready to use with Thermo Scientific™ Kits, Parts and Accessories for Orion™ 2118XP Oxygen Scavenger Analyzers. Choose from options including: oxygen scavenger consumables kit; reagent-less oxygen scavenger consumables kit; fluid connector block assembly with thumb screw and injection port; flow cell assembly with electrode holder, mounting bracket and O-rings; and mixing coil-tubing spool assembly.
6 x 1L iodide reagent and 6 x 6 ft. diffusion tubing assemblies with O-rings, reagent bottle adapter, 1 pint of 100ppm hydrazine standard, iodide electrode, reference electrode, 5 x 2 oz. reference electrode filling solution, O-ring kit, restriction tube
Electronics faceplate, air pump assembly, regulator assembly, 2 electrode cables with gland assemblies for sensing and reference electrodes, power supply assembly and fuse kit assembly
6 x 1L iodide reagent and 6 x 6 ft. diffusion tubing assemblies with O-rings, reagent bottle adapter, 1 pint of 100ppm hydrazine standard, iodide electrode, reference electrode, 5 x 2 oz. reference electrode filling solution, O-ring kit, restriction tube
Electronics faceplate, air pump assembly, regulator assembly, 2 electrode cables with gland assemblies for sensing and reference electrodes, power supply assembly and fuse kit assembly
Unit Size
Each
Showing 10 of 14Show more
Keep your oxygen scavenger analyzer ready to use with Thermo Scientific™ Kits, Parts and Accessories for Orion™ 2118XP Oxygen Scavenger Analyzers. Choose from options including: oxygen scavenger consumables kit; reagent-less oxygen scavenger consumables kit; fluid connector block assembly with thumb screw and injection port; flow cell assembly with electrode holder, mounting bracket and O-rings; and mixing coil-tubing spool assembly.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.