Biosate™ Peptone
Gibco™

Biosate™ Peptone

Biosate Peptone is an animal origin (AO) mixed hydrolysate comprised of 65% pancreatic digest of casein and 35% yeast extract.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
211862454 g
29431225 lb.
Catalog number 211862
Price (USD)
245.00
Each
Quantity:
454 g
Price (USD)
245.00
Each
Biosate Peptone is an animal origin (AO) mixed hydrolysate comprised of 65% pancreatic digest of casein and 35% yeast extract. This combination provides synergistic nutritional benefits that are not provided by the components alone, resulting in a unique nutritional profile. Customers have shown the combined use of these two peptones, when supplied as Biosate Peptone, improves toxin production with Clostridial species for vaccines, such as Clostridium botulinum and Clostridium perfringens.

Features:
• Hydrochloric acid hydrolysate of casein
• Animal origin nutritional supplement
• Milk protein rich in amino acid nitrogen

Applications of Biosate Peptone include:
• Intended for use as a nutritional supplement in vitamin assays, susceptibility testing, and other laboratory media, and microbial fermentation in which the high salt content will not interfere

Specifications
Certifications/ComplianceLUO, MUO
For Use With (Application)Nutritional supplement in vitamin assays, susceptibility testing, and other laboratory media, and microbial fermentation in which the high salt content will not interfere
FormatBottle
Product LineBiosate
Quantity454 g
Shelf Life1460 Days
SourceAnimal origin, casein and yeast
FormPowder
Product TypeDehydrated Culture Media
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.