Orion™ 2118XP Oxygen Scavenger Analyzer
Actual product may vary
Thermo Scientific™

Orion™ 2118XP Oxygen Scavenger Analyzer

Control spending and protect your system using the Thermo Scientific™ Orion™ 2118XP Oxygen Scavenger Analyzer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
2118XPO2 Scavenger Analyzer
2118XPENAnalyzer with Enclosure
Catalog number 2118XP
Price (USD)
-
Type:
O2 Scavenger Analyzer
Control spending and protect your system using the Thermo Scientific™ Orion™ 2118XP Oxygen Scavenger Analyzer. This analyzer offers unmatched results for the control and optimization of oxygen scavenger systems and provides the perfect balance of performance and ease of use.

Features:

  • Continuous online measurement ensures real time information for optimum control of the oxygen scavenger feed pump without excessive operating costs.
  • Measurements are fast and stable, limiting unnecessary calibration cycles due to drift with superior Orion iodide electrode technology.
  • Designed for versatility, our unique chemistry measures most oxygen scavengers with only a change in the standard.
  • Accurate and precise oxygen scavenger control maintains proper feed levels with confidence.
  • Repeatable and verifiable correlation of hydrazine readings to ELIMIN-OX concentration are obtained using a proprietary iodide reaction.

Ordering Information:

  • Annual Consumables Kit, Cat. No. 211850XP
  • Reagent, Cat. No. 181811
  • Diffusion Tubing, Cat. No. 181860
  • Calibration Kit, Cat. No. 2118XPCAL
  • Calibration Standards, Cat. No. 151810
  • Iodide Electrode, Cat. No. 100022
  • Reference Electrode, Cat. No. 100056
  • Reference Electrode Filling Solution, Cat. No. 181073
Specifications
Accuracy±5% or 2 ppb, whichever is greater
Accuracy (Temperature)±0.5°C
Accuracy (mV)± (0.5 mV + 0.1%)
Alarm Output3
Amperage5A
Analog Outputs2, Galvanically Isolated
CalibrationDouble Known Addition
Case MaterialValox 364
Certifications/ComplianceCE, CSA, UL, FCC Class A limits
Channel 1Hydrazine
Channel 2Optional ph/ORP or Conductivity
Channels1
Communication DetailsModbus with Additional Module
Current Loops2
Description2118XP Oxygen Scavenger Analyzer
Dimensions (L x W x H)11 x 17 x 26 in. (27 x 45 x 65cm)
Display TypeCustom backlit LCD with temperature, concentration, mV, error codes and menu driven prompts
Enclosure RatingWaterproof; IP66; NEMA 4X
Flow Rate40 mL/min.
Frequency0 to 60 Hz
Height (English)26 in.
Height (Metric)65 cm
Includes2118XP oxygen scavenger analyzer with iodide electrode, reference electrode, ATC probe, CD user guide and options kit
Inlet Pressure8 to 100 psig
Keypad TypeMembrane
Length (English)11 in.
Length (Metric)27 cm
Max. Relative Humidity5 to 95%, non-condensing
Measurement Ranges0 to 10 ppm
Mounting OptionsWall mount
Outputs0 to 20 mA; 4 to 20 mA
Relay Details3 Relay Outputs; Relay Load: 250VAC; 30VDC; 5A
Response Time95% within 2 minutes of injecting a standard solution
Sample Flow Rate40 mL/min nominal set by pressure regulator
Sample Inlet1/4 in. NPTF tube fitting
Sample Streams1
Temperature (English) Operating41°F to 113°F
Voltage85/132 VAC, 170/264 VAC
Width (English)17 in.
Width (Metric)45 cm
Temperature (Metric) Operating5°C to 35°C
TypeO2 Scavenger Analyzer
Unit SizeEach