Platinum(IV) oxide hydrate, typical Pt-content 79-84%
Platinum(IV) oxide hydrate, typical Pt-content 79-84%
Platinum(IV) oxide hydrate, typical Pt-content 79-84%
Platinum(IV) oxide hydrate, typical Pt-content 79-84%
Thermo Scientific Chemicals

Platinum(IV) oxide hydrate, typical Pt-content 79-84%

CAS: 52785-06-5 | O2Pt | 227.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
25 g
Packaging:
Glass bottle
Catalog number 211920250
Price (USD)
2,823.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
2,823.00
Each
Chemical Identifiers
CAS52785-06-5
IUPAC Namedioxoplatinum
Molecular FormulaO2Pt
InChI KeyYKIOKAURTKXMSB-UHFFFAOYSA-N
SMILESO=[Pt]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Appearance (Color)Brown to black
Surface area>=90 m²/g (BET)
Assay79 to 84 % (Pt)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only