Difco™ Tryptone Peptone
Actual product may vary
Difco™ Tryptone Peptone
Difco™ Tryptone Peptone
Gibco™

Difco™ Tryptone Peptone

Difco Tryptone Peptone is an animal origin (AO) pancreatic digest of casein. In addition to providing a unique nutritional profile, it has a higher nitrogen content than our other casein products. Tryptone Peptone supports the growth of many different types of bacteria and fungi, including fastidious organisms and in toxin production.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
2119225 lb.
2119211 lb.
21192325 lb.
Catalog number 211922
Price (USD)
758.00
Each
Quantity:
5 lb.
Price (USD)
758.00
Each

Difco Tryptone Peptone is an animal origin (AO) pancreatic digest of casein. In addition to providing a unique nutritional profile, it has a higher nitrogen content than our other casein products. Tryptone Peptone supports the growth of many different types of bacteria and fungi, including fastidious organisms and in toxin production. Customers have used this supplement in the development of animal health vaccines, such as Clostridial vaccines, and with human health Botox-like products produced from Clostridium botulinum.

Features:

  • Animal origin, pancreatic digest of casein
  • Primary nitrogen source in trypticase soy broth and agar
  • Referenced in Official Methods of Analysis of AOAC International
  • Supports the growth of many different types of bacteria and fungi

Applications of Tryptone Peptone include:

  • Recommended for use in media formulations when good growth of fungi and bacteria is required
  • Used in the development of animal health vaccines and with human health Botox-like products
Specifications
Certifications/ComplianceLUO, MUO
For Use With (Application)Dehydrated culture media
FormatPail
Quantity5 lb.
Shelf Life1460 Days
SourceAnimal Origin
FormPowder
Product TypeBacterial Media
Unit SizeEach
Contents & Storage
Store at 2°C to 25°C in cool, dry conditions in well-sealed containers.

Frequently asked questions (FAQs)

What alternative product do you recommend for Bacto Tryptone (Cat. No. 211699, 211701, 211705)?

We have several casein digests that could serve as alternatives to Bactone Tryptone (Cat. No. 211699, 211791, 211705), including Bacto Casitone (Cat. No. 225930, 225910), Difco Tryptone Peptone (Cat. No. 21921, 211922, 211923), BiTek Tryptone (Cat. No. 251420). However, the digestion and nutritive profile of each peptone are different, therefore there may be differences in bacterial growth or supplementation ability. We recommend empirically determining the performance of alternative casein digestion in your application.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.