Platinum(IV) oxide monohydrate, 99.8%, (trace metal basis)
Platinum(IV) oxide monohydrate, 99.8%, (trace metal basis)
Platinum(IV) oxide monohydrate, 99.8%, (trace metal basis)
Thermo Scientific Chemicals

Platinum(IV) oxide monohydrate, 99.8%, (trace metal basis)

CAS: 12137-21-2 | H2O3Pt | 245.10 g/mol
数量:
250 mg
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 211930010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS12137-21-2
IUPAC Namedioxoplatinum hydrate
Molecular FormulaH2O3Pt
InChI KeySPGAMGILENUIOF-UHFFFAOYSA-N
SMILESO.O=[Pt]=O
さらに表示
Platinum (Pt)77.6 to 82 %
Total trace metal impurities=<2000 ppm
Appearance (Form)Powder
Appearance (Color)Brown to black
Surface area>=80 m²/g
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only