Nuclear Fast Red, 94%, pure
Nuclear Fast Red, 94%, pure
Nuclear Fast Red, 94%, pure
Acros Organics

Nuclear Fast Red, 94%, pure

CAS: 6409-77-4 | C14H11NNaO7S | 360.292 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Glass bottle
Catalog number 211980050
HSN 98020000
Price (INR)
28,091.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
28,091.00
Each
Chemical Identifiers
CAS6409-77-4
IUPAC Namesodium 4-amino-1,3-dihydroxy-9,10-dioxo-9,10-dihydroanthracene-2-sulfonate
Molecular FormulaC14H8NNaO7S
InChI KeyIFSXZLJQEKGQAF-UHFFFAOYSA-M
SMILES[Na+].NC1=C(O)C(=C(O)C2=C1C(=O)C1=CC=CC=C1C2=O)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
HPLC>=92 %
Infrared spectrumConforms
Appearance (Color)Dark red to bordeaux
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only