Purpurin
Purpurin
Purpurin
Thermo Scientific Chemicals

Purpurin

CAS: 81-54-9 | C14H8O5 | 256.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 211990050
Price (USD)
41.65
Online Exclusive
45.60
Save 3.95 (9%)
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
41.65
Online Exclusive
45.60
Save 3.95 (9%)
Each
Chemical Identifiers
CAS81-54-9
IUPAC Name1,2,4-trihydroxy-9,10-dihydroanthracene-9,10-dione
Molecular FormulaC14H8O5
InChI KeyBBNQQADTFFCFGB-UHFFFAOYSA-N
SMILESOC1=CC(O)=C2C(=O)C3=CC=CC=C3C(=O)C2=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
UV Lambda max470 to 490 nm (10 mg/l methanol)
Appearance (Color)Dark red to orange-brown to brown
Infrared spectrumConforms
HPLC>=85 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only