Nalgene™ Large Wide-Mouth HDPE Bottles with Closure
Nalgene™ Large Wide-Mouth HDPE Bottles with Closure
Nalgene™ Large Wide-Mouth HDPE Bottles with Closure
Nalgene™ Large Wide-Mouth HDPE Bottles with Closure
Thermo Scientific™

Nalgene™ Large Wide-Mouth HDPE Bottles with Closure

Choose these large, wide-mouth HDPE bottles with closure for a wide range of lab and field uses, including dry chemical storage and water sampling.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)
2120-00104 L
2120-00052 L
Catalog number 2120-0010
Price (MXN)
-
Capacity (Metric):
4 L

Perform a wide range of lab and field applications including dry chemical storage and water sampling with Thermo Scientific™ Nalgene™ Large Wide-Mouth HDPE Bottles with Closure. These high-density, polyethylene bottles are good for use in the freezer to -100°C (-148°F). Chemical-resistant HDPE is compatible with a wide variety of lab reagents.

Features:
  • Excellent chemical resistance to most acids, bases and alcohols make this a versatile bottle
  • Can be used in freezer to -100°C (-148°F) for long-term storage
  • Linerless cap closes the bottle without a liner that can wrinkle, cause leaks and contaminate your solutions
  • This bottle is not liquid leakproof†
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials

Includes:

100 mm polypropylene screw caps.

Recycling Information:

Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
Capacity1 gal., 4 L
Capacity (English)1 gal.
Certifications/ComplianceManufactured in compliance with ISO 13485:2016 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Closure ColorWhite
Closure LinerUnlined
Closure MaterialPolypropylene
Closure Size100 mm
Closure TypeScrew Cap
DescriptionLarge Wide-Mouth Bottle
Diameter (Metric)153 mm
Diameter (Metric) Inner Neck89 mm
DisposableReusable
Height (English)10.98 in.
Height (Metric)288 mm
Neck Finish100 mm
No. per Case6/Cs.
No. per Pack1/Pk.
Product TypeWide Mouth Bottle
Quantity1/Pk., 6/Cs.
ShapeRound
Through the Closure DispensingNo
Width (English)5.02 in.
Capacity (Metric)4 L
Closure IncludedYes
ColorNatural
GraduatedNo
MaterialHDPE
MouthWide
SterilityNon-sterile
Unit SizeCase of 6

Frequently asked questions (FAQs)

Why are some of your wide mouth bottles and jars not considered liquid leakproof?

This was a consequence of design and engineering limitations, and there was no practical way to incorporate the same level of seal integrity as our certified leakproof containers. While in many cases, they still hold up well to liquid containment applications, they have not been tested specifically for that purpose, and thus would be the responsibility of the customer to perform validation and risk management studies if necessary.