Product Image
Thermo Scientific™
Orion™ 2120XP Calcium Hardness Analyzer Accessories and Kits
Accessories and kits for the Thermo Scientific™ Orion™ 2120XP Calcium Hardness Analyzer include a consumables kit, calibration kit, carrying case, electrode kit and faceplate.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
212050XPCa Hardness Consumables Kit
212050XPFReagentless CA Hardness Kit
2120XPCALCalibration Kit
2120XPEKElectrode Kit
2120XPEPElectronics Faceplate
Catalog number 212050XP
Price (USD)
2,394.00
Online exclusive
2,660.00
Save 266.00
Each
Type:
Ca Hardness Consumables Kit
Price (USD)
2,394.00
Online exclusive
2,660.00
Save 266.00
Each
Use the Thermo Scientific™ Orion™ 2120XP Calcium Hardness Analyzer Accessories and Kits with the Thermo Scientific™ Orion™ 2120XP Calcium Hardness Analyzer. Accessories and kits include a calcium consumables kit, a reagent-less calcium hardness kit, a calcium hardness calibration kit with carrying case, a calcium electrode kit or a electronics faceplate.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Certifications/ComplianceCE, CSA, UL, FCC Class A limits
DescriptionCalcium Hardness Consumables Kit - for one year operation, includes reagent and diffusion tubing, calibration standards, sensing electrode and reference electrode with filling solution
For Use With (Equipment)Orion™ 2120XP Calcium Hardness Analyzer
Includes6 x 1L formic acid reagent and 6 x 2 ft. diffusion tubing with O-rings (182011), calcium calibration standards kit (182040), calcium electrode (100049), reference electrode (100056), 5 x 2 oz. reference electrode filling solution (181073), O-ring kit (2100OK), check valve (2100VC), box of pipette tips (204846-001), restriction tube assembly (2100RT) and inlet filter with gaskets (181170)
Temperature (Metric) Operating5°C to 45°C (Ambient)
TypeCa Hardness Consumables Kit
Unit SizeEach