New Fuchsin
New Fuchsin
New Fuchsin
Thermo Scientific Chemicals

New Fuchsin

CAS: 3248-91-7 | C22H24ClN3 | 365.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 212141000
Price (USD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS3248-91-7
IUPAC Nametris(4-amino-3-methylphenyl)methylium chloride
Molecular FormulaC22H24ClN3
InChI KeyORDGVBRMUJCHEO-UHFFFAOYSA-M
SMILES[Cl-].CC1=CC(=CC=C1N)[C+](C1=CC=C(N)C(C)=C1)C1=CC=C(N)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
UV Lambda max550 to 554 nm (in 50% ethanol)
Appearance (Form)Crystalline powder or crystals
Specific absorbance A (1%/1cm)>=1500 (at lambda max.)
Infrared spectrumConforms
Absorption Ratio P-15/P+151.1 to 1.4
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only