Luxol™ Fast Blue MBSN, pure
Luxol™ Fast Blue MBSN, pure
Luxol™ Fast Blue MBSN, pure
Thermo Scientific Chemicals

Luxol™ Fast Blue MBSN, pure

CAS: 1328-51-4 | C32H14CuN8Na2O6S2 | 780.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 212171000
Price (USD)
162.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
162.00
Each
Chemical Identifiers
CAS1328-51-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
UV spectrumComplies
Infrared spectrumConforms
Appearance (Color)Blue
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only