Naphthol AS, 99%
Naphthol AS, 99%
Naphthol AS, 99%
Thermo Scientific Chemicals

Naphthol AS, 99%

CAS: 92-77-3 | C17H13NO2 | 263.28 g/mol
Have Questions?
Catalog NumberQuantityPackaging
212181000100 gGlass bottle
Catalog number 212181000
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.90
Save 8.25
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
47.65
Special offer
Online exclusive
Ends: 28-Dec-2026
55.90
Save 8.25
Each
Chemical Identifiers
CAS92-77-3
IUPAC Name3-hydroxy-N-phenylnaphthalene-2-carboxamide
Molecular FormulaC17H13NO2
InChI KeyJFGQHAHJWJBOPD-UHFFFAOYSA-N
SMILESOC1=C(C=C2C=CC=CC2=C1)C(=O)NC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
UV spectrumComplies
Water=<0.3 %
Appearance (Color)Beige
Melting point>=243°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only