Aluminum oxide, 99.99%, (trace metal basis), extra pure
Aluminum oxide, 99.99%, (trace metal basis), extra pure
Aluminum oxide, 99.99%, (trace metal basis), extra pure
Thermo Scientific Chemicals

Aluminum oxide, 99.99%, (trace metal basis), extra pure

CAS: 1344-28-1 | Al2O3 | 101.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
Glass bottle
Catalog number 212251000
Price (USD)
273.65
Online Exclusive
304.00
Save 30.35 (10%)
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
273.65
Online Exclusive
304.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS1344-28-1
IUPAC Namedialuminium(3+) trioxidandiide
Molecular FormulaAl2O3
InChI KeyPNEYBMLMFCGWSK-UHFFFAOYSA-N
SMILES[O--].[O--].[O--].[Al+3].[Al+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Total trace metal impurities=<0.01 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only