Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure
Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure
Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure
Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure
Thermo Scientific™

Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure

Collect, store and transport dry chemicals and environmental samples with this large square wide-mouth HDPE bottle.
Have Questions?
Catalog NumberCapacity (Metric)
2123-0010PK
also known as 2123-0010
4 L
Catalog number 2123-0010PK
also known as 2123-0010
Price (USD)
-
Capacity (Metric):
4 L

Collect, store and transport dry chemicals and environmental samples with Thermo Scientific™ Nalgene™ Large Square Wide-Mouth HDPE Bottle with Closure. Molded-in handgrips provide greater convenience and safety when pouring.

  • Useful for collecting, storing and transporting dry chemicals, environmental samples, reagents and distilled water
  • Good for freezing to -100°C (-148°F)
  • Excellent chemical resistance to most acids, bases and alcohols
  • Square shape saves space, handles easily
  • Wide mouth gives easy access
  • Polypropylene cap included
  • Not liquid leakproof
  • Bottle and Carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.

Specifications
Capacity (English)1 gal.
Certifications/ComplianceUSP Class VI; FDA - 21 CFR
Closure ColorWhite
Closure MaterialPolypropylene
Closure Size100 mm
Closure TypeScrew Cap
DescriptionLarge Wide-Mouth Square Bottle
Diameter (English)5.6 in.
Diameter (English) Inner Neck3.5 in.
Diameter (Metric)142 mm
Diameter (Metric) Inner Neck89 mm
Height (English)11.5 in.
Height (Metric)292 mm
No. per Pack1
Product TypeWide Mouth Bottle (Large)
ShapeSquare
Through the Closure DispensingNo
Width (English)5.66 in.
Capacity (Metric)4 L
Closure IncludedYes
ColorTranslucent
GraduatedNo
MaterialHDPE
MouthWide
SterilityNon-sterile
Unit SizeEach

Frequently asked questions (FAQs)

Why are some of your wide mouth bottles and jars not considered liquid leakproof?

This was a consequence of design and engineering limitations, and there was no practical way to incorporate the same level of seal integrity as our certified leakproof containers. While in many cases, they still hold up well to liquid containment applications, they have not been tested specifically for that purpose, and thus would be the responsibility of the customer to perform validation and risk management studies if necessary.