Difco™ Soytone
Gibco™

Difco™ Soytone

Difco Soytone is an enzymatic digest of soy. With high concentrations of vitamins and carbohydrates uniquely present in soy, thisRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
21248910 kg
212488500 g
Catalog number 212489
Price (BRL)
10.530,52
Each
Quantity:
10 kg
Price (BRL)
10.530,52
Each
Difco Soytone is an enzymatic digest of soy. With high concentrations of vitamins and carbohydrates uniquely present in soy, this supplement offers an alternative carbohydrate profile to yeast supplements for mammalian cell processes, such as CHO.

Features:
• An enzymatic digest of soy
• High concentrations of vitamins and carbohydrate

Applications of Difco Soytone include:
• An alternative carbohydrate profile to yeast supplements for mammalian cell processes, such as CHO

Specifications
Culture TypeMammalian, Insect, Microbial
For Use With (Application)Mammalian Cell Culture
Product LineDifco
Quantity10 kg
SourceAnimal origin-free
FormPowder
Product TypeYeast Supplement
Unit SizeEach
Contents & Storage
Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.