Iodine, 99.5+%, for analysis
Iodine, 99.5+%, for analysis
Iodine, 99.5+%, for analysis
Thermo Scientific Chemicals

Iodine, 99.5+%, for analysis

C-X bond formation reagent | CAS: 7553-56-2 | I2 | 253.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 212495000
Price (TWD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS7553-56-2
IUPAC Namediiodine
Molecular FormulaI2
InChI KeyPNDPGZBMCMUPRI-UHFFFAOYSA-N
SMILESII
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Glistening spheres
Insoluble matterPasses test
Calcium (Ca)=<10 ppm
Iron (Fe)=<50 ppm
Sodium (Na)=<50 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only