Lead(II) carbonate, ACS reagent
Lead(II) carbonate, ACS reagent
Lead(II) carbonate, ACS reagent
Thermo Scientific Chemicals

Lead(II) carbonate, ACS reagent

CAS: 598-63-0 | CO3Pb | 267.20 g/mol
製品番号(カタログ番号) 212515000
価格(JPY)
-
数量:
500 g
一括またはカスタム形式をリクエストする
化学物質識別子
CAS598-63-0
IUPAC Nameλ²-lead(2+) carbonate
Molecular FormulaCO3Pb
InChI KeyMFEVGQHCNVXMER-UHFFFAOYSA-L
SMILES[Pb++].[O-]C([O-])=O
さらに表示
Appearance (Color)White
Chloride (Cl)=<0.002 %
Calcium (Ca)=<0.01 %
Potassium (K)=<0.02 %
Nitrate and Nitrite (as NO3)(limit about 0.005%) Passes test
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only