Lithium nitrate, 99.999%, (trace metal basis)
Lithium nitrate, 99.999%, (trace metal basis)
Lithium nitrate, 99.999%, (trace metal basis)
Thermo Scientific Chemicals

Lithium nitrate, 99.999%, (trace metal basis)

CAS: 7790-69-4 | LiNO3 | 68.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 212540050
Price (TWD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7790-69-4
IUPAC Namelithium(1+) nitrate
Molecular FormulaLiNO3
InChI KeyIIPYXGDZVMZOAP-UHFFFAOYSA-N
SMILES[Li+].[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder and/or chunks
Appearance (Color)White
Lithium (Li)9 to 11 %
Total trace metal impurities=<10 ppm
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for ultra trace analysis
RUO – Research Use Only