Ammonium titanyl oxalate monohydrate, 98%, pure
Ammonium titanyl oxalate monohydrate, 98%, pure
Ammonium titanyl oxalate monohydrate, 98%, pure
Ammonium titanyl oxalate monohydrate, 98%, pure
Thermo Scientific Chemicals

Ammonium titanyl oxalate monohydrate, 98%, pure

CAS: 10580-03-7 | C4H8N2O9Ti | 275.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 212730100
Price (USD)
84.65
Special offer
Online exclusive
Ends: 28-Dec-2026
99.00
Save 14.35
Each
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
84.65
Special offer
Online exclusive
Ends: 28-Dec-2026
99.00
Save 14.35
Each
Chemical Identifiers
CAS10580-03-7
IUPAC Namediammonium oxotitaniumbis(ylium) dioxalate
Molecular FormulaC4H8N2O9Ti
InChI KeyVBUNWMBOVJSSMH-UHFFFAOYSA-L
SMILES[NH4+].[NH4+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light beige
Appearance (Form)Crystalline powder or crystals
Infrared spectrumConforms
Titration with KMnO497.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only