Zinc bromide, 99.999%, (trace metal basis), extra pure
Zinc bromide, 99.999%, (trace metal basis), extra pure
Zinc bromide, 99.999%, (trace metal basis), extra pure
Thermo Scientific Chemicals

Zinc bromide, 99.999%, (trace metal basis), extra pure

CAS: 7699-45-8 | Br2Zn | 225.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
Catalog number 212770100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7699-45-8
IUPAC Namezinc(2+) dibromide
Molecular FormulaBr2Zn
InChI KeyVNDYJBBGRKZCSX-UHFFFAOYSA-L
SMILES[Zn++].[Br-].[Br-]
View more
SpecificationsSpecification SheetSpecification Sheet
Total trace metal impurities=<0.001 %
Appearance (Form)Crystalline powder
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysis
RUO – Research Use Only