Bromuro de cinc, 99,999 %, (base de trazas de metales), extra puro, Thermo Scientific Chemicals
Bromuro de cinc, 99,999 %, (base de trazas de metales), extra puro, Thermo Scientific Chemicals
Bromuro de cinc, 99,999 %, (base de trazas de metales), extra puro, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bromuro de cinc, 99,999 %, (base de trazas de metales), extra puro, Thermo Scientific Chemicals

CAS: 7699-45-8 | Br2Zn | 225.19 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
2.5 g
50 g
Envase:
Frasco de vidrio
Número de catálogo 212770500
Precio (USD)
-
Cantidad:
50 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS7699-45-8
IUPAC Namezinc(2+) dibromide
Molecular FormulaBr2Zn
InChI KeyVNDYJBBGRKZCSX-UHFFFAOYSA-L
SMILES[Zn++].[Br-].[Br-]
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Crystalline powder
Total trace metal impurities=<0.001 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Para análisis de ultratrazas
RUO – Research Use Only