Tetrabutylammonium borohydride, 98%
Tetrabutylammonium borohydride, 98%
Tetrabutylammonium borohydride, 98%
Thermo Scientific Chemicals

Tetrabutylammonium borohydride, 98%

CAS: 33725-74-5 | C16H40BN | 257.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 212900500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS33725-74-5
IUPAC Nametetrabutylazanium boranuide
Molecular FormulaC16H40BN
InChI KeyGMBOFJFPOCGSOI-UHFFFAOYSA-N
SMILES[BH4-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals and/or chunks
Appearance (Color)White to off-white
Titration with HClO4>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only