Tetrabutylammonium hydroxide, 1M solution in methanol
Tetrabutylammonium hydroxide, 1M solution in methanol
Tetrabutylammonium hydroxide, 1M solution in methanol
Thermo Scientific Chemicals

Tetrabutylammonium hydroxide, 1M solution in methanol

CAS: 2052-49-5 | C16H37NO | 259.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
2.5 L
Catalog number 212910025
Price (USD)
913.65
Special offer
Online exclusive
Ends: 28-Dec-2026
1,075.00
Save 161.35
Each
Quantity:
2.5 L
Request bulk or custom format
Price (USD)
913.65
Special offer
Online exclusive
Ends: 28-Dec-2026
1,075.00
Save 161.35
Each
Chemical Identifiers
CAS2052-49-5
IUPAC Nametetrabutylazanium hydroxide
Molecular FormulaC16H37NO
InChI KeyVDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES[OH-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
Titration with HCl0.95 to 1.10 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only