Cobalt(II) nitrate hexahydrate, 99%, pure
Cobalt(II) nitrate hexahydrate, 99%, pure
Cobalt(II) nitrate hexahydrate, 99%, pure
Thermo Scientific Chemicals

Cobalt(II) nitrate hexahydrate, 99%, pure

CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic bottle
Catalog number 213090010
Price (JPY)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS10026-22-9
IUPAC Nameλ²-cobalt(2+) hexahydrate dinitrate
Molecular FormulaCoH12N2O12
InChI KeyQGUAJWGNOXCYJF-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering crystalline powder or crystals or chips
Appearance (Color)Pink to red to brown
Titration Complexometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only