Magnesium sulfate heptahydrate, 99.5%, for analysis
Magnesium sulfate heptahydrate, 99.5%, for analysis
Magnesium sulfate heptahydrate, 99.5%, for analysis
Thermo Scientific Chemicals

Magnesium sulfate heptahydrate, 99.5%, for analysis

CAS: 10034-99-8 | H14MgO11S | 246.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
25 kg
Packaging:
View all
Plastic bottle
Catalog number 213115000
Price (USD)
118.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
118.00
Each
Chemical Identifiers
CAS10034-99-8
IUPAC Namemagnesium(2+) heptahydrate sulfate
Molecular FormulaH14MgO11S
InChI KeyWRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<5 ppm
Heavy metals (as Pb)=<5 ppm
Potassium (K)=<50 ppm
Appearance (Form)Crystals
Insoluble matter=<0.005 % (in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only