Thioflavin S
Thermo Scientific Chemicals

Thioflavin S

CAS: 1326-12-1
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 213150050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1326-12-1
SpecificationsSpecification SheetSpecification Sheet
Solubility(0.1 % in water/EtOH (1:1)) Clear to hazy
Infrared spectrumConforms
UV Lambda max360 to 380 nm (in water)
Appearance (Color)Tan to brown
UV spectrumComplies
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only