1-Hexene, 97%
1-Hexene, 97%
1-Hexene, 97%
Thermo Scientific Chemicals

1-Hexene, 97%

1-Hexene, 97%, C6H12, CAS Number-592-41-6, dialene 6, butyl ethylene, 1-hexene, butylethylene, 1-n-hexene, hexylene, hexene-1, hexene, n-hexene, unii-b38zz8c206, 1L, 96% min. (GC), 01, 215, 62.0 deg.C to 65.0 deg.C, 100.0, 96.0, CHEBI:24579, Colorless, 0.6780g/mL, 209-753-1, -26 deg.C, 84.15 | CAS: 592-41-6 | C6H12 | 84.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
2.5 L
25 L
Packaging:
View all
Metal Drum
Catalog number 213210250
Price (USD)
-
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Chemical Identifiers
CAS592-41-6
IUPAC Namehex-1-ene
Molecular FormulaC6H12
InChI KeyLIKMAJRDDDTEIG-UHFFFAOYSA-N
SMILESCCCCC=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=96.0 %
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.3867 to 1.3887 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only