EZ-Link™ NHS-LC-Biotin - FAQs
View additional product information for EZ-Link™ NHS-LC-Biotin - FAQs (21336)
2 product FAQs found
What should I do if my EZ-Link NHS-LC-Biotin (Cat. No. 21336) is found crystalized?
The Z-Link NHS-LC-Biotin (Cat. No. 21336) should be a powder but not in crystalized form. So, in that case it is hydrolyzed and should not be used as wouldn't be reliable any more.
Find additional tips, troubleshooting help, and resources within our Labeling Chemistry Support Center.
Can you provide the shelf-life for EZ-Link NHS-LC-Biotin?
EZ-Link NHS-LC-Biotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (4 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.