EZ-Link™ Sulfo-NHS-LC-LC-Biotin, 50 mg - FAQs
View additional product information for EZ-Link™ Sulfo NHS-LC-LC-Biotin, No-Weigh™ Format - FAQs (A35358, 21338)
4 product FAQs found
Can you provide the shelf-life for EZ-Link Sulfo-NHS-LC-LC-Biotin?
EZ-Link Sulfo-NHS-LC-LC-Biotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What is the difference between EZ-Link Sulfo-NHS-LC-Biotin and Sulfo-NHS-LC-LC-Biotin?
EZ-Link Sulfo-NHS-LC-Biotin has a spacer arm length of 22.4 Angstroms while EZ-Link Sulfo-NHS-LC-LC-Biotin has a spacer arm length of 30.5 Angstroms.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Is there any advantage of choosing a Sulfo-NHS biotin with a longer spacer arm than EZ-Link Sulfo NHS Biotin?
Sometimes. The biotin has to insert 9 angstrom into the pocket of avidin, streptavidin or NeutrAvidin to bind. If steric hindrance may be a problem, using a biotinylation reagent with a longer spacer arm (Sulfo-NHS-LC Biotin or Sulfo-NHS-LC-LC Biotin) may result in greater purification or deteciton.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Why would I chose a Sulfo-NHS Biotin over its non-sulfonated counterpart?
The sodium sulfonate group lends water solubility to the reagent.
Note: The actual biotin moiety that is added is identical between NHS and Sulfo-NHS biotins that vary only in the addition of the sodium sulfonate on the leaving group. Researchers preferring a more water soluble biotin be added to their molecule should consider a PEGylated version of biotin.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.