Medium NCTC-109 (1X)
Gibco™

Medium NCTC-109 (1X)

NCTC-109 medium is a chemically defined medium that was originally developed by the National Cancer Institute and formulated for theRead more
Have Questions?
Catalog NumberQuantity
21340039500 mL
Catalog number 21340039
Price (MXN)
-
Quantity:
500 mL
Customize this product
NCTC-109 medium is a chemically defined medium that was originally developed by the National Cancer Institute and formulated for the culture of mouse Strain L sublines, NCTC clone 929 (L929 cells) in a serum-free environment. Gibco™ NCTC-109 medium is also suitable for generating and maintaining hybridoma cells.
This NCTC-109 is modified as follows:
WithWithout
• Phenol Red• L-glutamine
• Coenzymes• HEPES

The complete formulation is available.

Gibco™ NCTC-109 medium is rich in nucleosides, vitamins, coenzymes, reducing agents and other unique ingredients.

Product Intended Use
For in vitro diagnostic use. CAUTION: Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.

cGMP Manufacturing and Quality System
Gibco™ NCTC-109 medium is manufactured at a cGMP compliant facility, located in Grand Island, New York. The facility is registered with the FDA as a medical device manufacturer and is certified to ISO 13485 standards. For supply chain continuity, we offer an identical Gibco™ NCTC-109 product made in our Scotland facility (21340-054). This facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.

NCTC-109 contains no proteins, lipids, or growth factors. NCTC-109 medium typically requires supplementation of a growth factor cocktail (for serum free growth) or Fetal Bovine Serum (FBS). NCTC-109 medium uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.
For in vitro diagnostic use. Not for human or animal therapeutic use. Uses other than the labeled intended use may be a violation of local law.
Specifications
Cell TypeHybridomas
Product LineGibco
Product TypeCell Culture Medium
Quantity500 mL
Shipping ConditionRoom Temperature
FormLiquid
With AdditivesPhenol Red
Without AdditivesNo Glutamine, No HEPES, No Sodium Pyruvate
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 24 months from date of manufacture