Calcium phosphate, for analysis, 35-40% (Ca), mixture of Calcium phosphates
Calcium phosphate, for analysis, 35-40% (Ca), mixture of Calcium phosphates
Calcium phosphate, for analysis, 35-40% (Ca), mixture of Calcium phosphates
Thermo Scientific Chemicals

Calcium phosphate, for analysis, 35-40% (Ca), mixture of Calcium phosphates

CAS: 7758-87-4 | Ca3O8P2 | 310.18 g/mol
製品番号(カタログ番号) 213480050
価格(JPY)
-
数量:
5 kg
一括またはカスタム形式をリクエストする
化学物質識別子
CAS7758-87-4
IUPAC Nametricalcium diphosphate
Molecular FormulaCa3O8P2
InChI KeyQORWJWZARLRLPR-UHFFFAOYSA-H
SMILES[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O
さらに表示
Arsenic (As)=<0.0003 %
Barium (Ba)Passes test
Iron (Fe)=<0.04 %
Appearance (Color)White
Appearance (Form)Powder
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only