Stains-all, 97%, pure
Stains-all, 97%, pure
Stains-all, 97%, pure
Thermo Scientific Chemicals

Stains-all, 97%, pure

CAS: 7423-31-6 | C30H28BrN2S2+ | 560.592 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 213510050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7423-31-6
IUPAC Name1-ethyl-2-{3-[(2Z)-1-ethyl-1H,2H-naphtho[1,2-d][1,3]thiazol-2-ylidene]-2-methylprop-1-en-1-yl}naphtho[1,2-d][1,3]thiazol-1-ium bromide
Molecular FormulaC30H27BrN2S2
InChI KeyMPBRYMWMMKKRGC-UHFFFAOYSA-M
SMILES[Br-].CCN1\C(SC2=CC=C3C=CC=CC3=C12)=C\C(C)=CC1=[N+](CC)C2=C(S1)C=CC1=CC=CC=C21
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
UV spectrumComplies
TLC>=96.0 %
Appearance (Color)Dark grey to dark purple
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only