MagnaBind™ Magnet for 96-Well Plate Separator
Catalog number: 21358
Thermo Scientific™  Related applications: Protein Assays and Analysis | Protein Purification & Isolation
Error loading your content!
  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons
Your on-site stock
›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top
Description
Thermo Scientific MagnaBind Magnet for 96-Well Plate Separator, formulation: plastic holder containing strong, rare-earth magnet. Sufficient for collecting magnetic particles in wells of standard 96-well microplates.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment): MagnaBind™ Magnetic Beads and Magnets
Product Line: MagnaBind™
Product Size: 1 magnet
Type: Base
Contents & storage
0.0
Documents
Manuals & protocols