EZ-Link™ NHS-PEG4-Biotin, 50 mg - FAQs
View additional product information for EZ-Link™ NHS-PEG4-Biotin - FAQs (21330, 21363, 21362, A39259)
1 product FAQs found
Can you provide the shelf-life for EZ-Link NHS-PEG4-Biotin?
EZ-Link NHS-PEG4-Biotin is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.