Thermo Scientific Chemicals

Molybdic acid, for analysis, Thermo Scientific Chemicals

Catalog number: 213625000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Molybdic acid, for analysis, Thermo Scientific Chemicals

Catalog number: 213625000
500 g, Plastic bottle, Each
Quantity
Packaging

Chemical Identifiers

CAS
7782-91-4
IUPAC Name
dioxomolybdenumdiol
Molecular Formula
H2MoO4
InChI Key
VLAPMBHFAWRUQP-UHFFFAOYSA-L
SMILES
O[Mo](O)(=O)=O
Appearance (Color)
White to light yellow
Additional info
contains ammonium molybdate
Appearance (Form)
Powder
Titration with NaOH
>=85 % (as MoO3)
Heavy metals (as Pb)
=<30 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text